Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zirconium(IV) oxide (ZrO₂:TiO₂ - 90:10)% (w/w) stabilised, tablets

Supplier: Alfa Aesar
Warning
Synonyms: Zirconium dioxideZirconium Oxide

Stabilisation: Stabilised with Titanium (10% TiO₂)

45126.22EA 138 EUR InStock
45126.22 45126.06 45126.14
Zirconium(IV) oxide (ZrO₂:TiO₂ - 90:10)% (w/w) stabilised, tablets
Zirconium (IV) oxide


Formula: ZrO₂
MW: 123,22 g/mol
Boiling Pt: 4300 °C (1013 hPa)
Melting Pt: ∼2500 °C
Density: 5,4
Storage Temperature: Ambient
MDL Number: MFCD00011310
CAS Number: 1314-23-4
EINECS: 215-227-2
UN: 0000
Merck Index: 14,10180
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty