4-Chloro-2-phenylquinazoline

MAYBRH01997.1EA 0 EUR
CHMPRH01997.250 MAYBRH01997.1
4-Chloro-2-phenylquinazoline
4-Chloro-2-phenylquinazoline
Formula: C₁₄H₉ClN₂
MDL Number: MFCD00006713
CAS Number: 6484-25-9
EINECS: 229-346-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR