Isosorbide dimethyl ether

Supplier: LGC Standards PROMOCHEM

PROOMM0126.02EA 0 EUR
PROOMM0126.02
Isosorbide dimethyl ether
Isosorbide dimethyl ether
Formula: C₈H₁₄O₄
MW: 174,20
MDL Number: MFCD00064828
CAS Number: 5306-85-4
EINECS: 226-159-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR