1-(2-Furoyl)piperazine ≥98%

Supplier: Thermo Scientific

Warning

A16773.22EA 79.4 EUR
A16773.22 A16773.36
1-(2-Furoyl)piperazine ≥98%
1-(2-Furoyl)piperazine
Formula: C₉H₁₂N₂O₂
MW: 180,21
Boiling Pt: 137…140 °C (0,1 mmHg)
Melting Pt: 66…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00038831
CAS Number: 40172-95-0
EINECS: 254-823-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR