Cyclohexyldiphenylphosphine 98%
|
Warning
|
Synonyms:
Diphenylcyclohexylphosphine
ACRO406100050EA
99.5
EUR
ACRO406100050
ACRO406100010
Cyclohexyldiphenylphosphine 98%
Cyclohexyldiphenylphosphine
Formula:
C₁₈H₂₁P MW: 268,33 g/mol Melting Pt: 53 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00046360 CAS Number: 6372-42-5 EINECS: 228-904-2 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥97.5 % |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...