TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)

Supplier: Molekula

Synonyms: TBTU

MOLE39093565-5GEA 0 EUR
MOLE39093565-5G
TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)
TBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)
Formula: C₁₁H₁₆BF₄N₅O
MW: 321,1 g/mol
MDL Number: MFCD00077413
CAS Number: 125700-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR