2-Amino-5-methyl-4-phenylthiazole, 97%

H51825.06EA 83.8 EUR
ALFAH51825.06 ALFAH51825.03
2-Amino-5-methyl-4-phenylthiazole, 97%
Purity: 97%
CAS No.: 30709-67-2
Molecular Formula: C10H10N2S
Formula Weight: 190.26
Melting Point: 107-112°
MDL No.: MFCD00617257
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR