Ethyl-4-methyl-2-(2-pyridinyl)-1,3-thiazole-5-carboxylate ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Ethyl 4-methyl-2-(2-pyridyl)thiazole-5-carboxylate
H54842.03EA
49.9
EUR
H54842.03
Ethyl-4-methyl-2-(2-pyridinyl)-1,3-thiazole-5-carboxylate ≥97%
Ethyl 4-methyl-2-(2-pyridinyl)-1,3-thiazole-5-carboxylate
Formula:
C₁₂H₁₂N₂O₂S MW: 248,30 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01936157 CAS Number: 383146-88-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...