4-Methoxystilbene ≥98%

Supplier: Thermo Scientific

A17328.06EA 77 EUR
A17328.06 A17328.14 A17328.22
4-Methoxystilbene ≥98%
4-Methoxystilbene
Formula: C₁₅H₁₄O
MW: 210,27
Melting Pt: 136…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00017178
CAS Number: 1142-15-0
EINECS: 214-530-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR