1-Amino-2,4-dibromo-9,10-anthraquinone ≥97%
|
Warning
|
Synonyms:
1-Amino-2,4-dibromoanthraquinone
ABCRAB124984.0250
1-Amino-2,4-dibromo-9,10-anthraquinone ≥97%
1-Amino-2,4-dibromo-9,10-anthraquinone
Formula:
C₁₄H₇Br₂NO₂ MW: 381,03 g/mol Melting Pt: 225…229 °C Storage Temperature: Ambient |
MDL Number:
MFCD00019155 CAS Number: 81-49-2 EINECS: 201-354-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...