1,4-Bis(methylamino)-9,10-anthraquinone 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1,4-Bis(methylamino)anthraquinone,Disperse Blue 14,1,4-Bis(methylamino)anthracene-9,10-dione
L00903.06
L00903.06EA
53.5
EUR
L00903.06
1,4-Bis(methylamino)-9,10-anthraquinone 95%
1,4-Bis(methylamino)-9,10-anthraquinone
Formula:
C₁₆H₁₄N₂O₂ MW: 266,29 g/mol Melting Pt: ∼ 215 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00001198 CAS Number: 2475-44-7 EINECS: 219-602-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...