6-Fluorochromone-2-carboxylic acid ≥97%

Warning

Synonyms: 6-Fluoro-4-oxo-4H-chromene-2-carboxylic acid

H32714.03EA 79.1 EUR
H32714.03
6-Fluorochromone-2-carboxylic acid ≥97%
6-Fluorochromone-2-carboxylic acid
Formula: C₁₀H₅FO₄
MW: 208,15
Melting Pt: 257…259 °C
Storage Temperature: Ambient
MDL Number: MFCD03070543
CAS Number: 99199-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR