Dicyclohexylchlorophosphine 97%
|
Danger
|
Synonyms:
Chlorodicyclohexylphosphine
ACRO353290010EA
72.9
EUR
ACRO353290010
ACRO353290050
Dicyclohexylchlorophosphine 97%
Dicyclohexylchlorophosphine
Formula:
C₁₂H₂₂ClP MW: 232,73 g/mol Boiling Pt: 165 °C (12 mmHg) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014294 CAS Number: 16523-54-9 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5310 to 1.5340 (20°C, 589 nm) |
Titration Argentometric | ≥96.0 % (ex Cl) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...