Ethyl-3-amino-1H-pyrazole-4-carboxylate 99%
Supplier: Thermo Scientific
Synonyms:
3-Amino-4-carbethoxypyrazole
, Ethyl 3-amino-1H-pyrazole-4-carboxylate
ACRO103310050EA
57
EUR
ACRO103310050
Ethyl-3-amino-1H-pyrazole-4-carboxylate 99%
Ethyl 3-amino-1H-pyrazole-4-carboxylate
Formula:
C₆H₉N₃O₂ MW: 155,16 g/mol Melting Pt: 103…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005238 CAS Number: 6994-25-8 EINECS: 230-262-3 |
Specification Test Results
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 103 to 107 °C |
HPLC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...