3,5-Dimethyl-1H-pyrazole-1-carboxamide ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1-Carbamoyl-3,5-dimethylpyrazole
, 3,5-Dimethylpyrazole-1-carboxamide
A14523.09EA
26.9
EUR
A14523.09
3,5-Dimethyl-1H-pyrazole-1-carboxamide ≥98%
3,5-Dimethyl-1H-pyrazole-1-carboxamide
Formula:
C₆H₉N₃O MW: 139,16 g/mol Melting Pt: 112…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003126 CAS Number: 934-48-5 EINECS: 213-283-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...