3-Amino-5-phenyl-1H-pyrazole 98%

Supplier: Thermo Scientific

Warning

Synonyms: 3-Amino-5-phenylpyrazole

ACRO368020050EA 216 EUR
ACRO368020050 ACRO368020010
3-Amino-5-phenyl-1H-pyrazole 98%
3-Amino-5-phenyl-1H-pyrazole
Formula: C₉H₉N₃
MW: 159,19
Melting Pt: 118…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00191749
CAS Number: 1572-10-7

Order Now

Specification Test Results

Appearance Off-white to brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 124 to 129 °C
GC ≥97.5 %
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR