2-Benzylamino-4-benzyloxy-5-bromopyrimidine 95%

Supplier: Thermo Scientific
Warning

H27372.MDEA 79.5 EUR
H27372.MD H27372.03
2-Benzylamino-4-benzyloxy-5-bromopyrimidine 95%
2-Benzylamino-4-benzyloxy-5-bromopyrimidine
Formula: C₁₈H₁₆BrN₃O
Storage Temperature: Ambient
MDL Number: MFCD06798242
CAS Number: 885952-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR