2-Benzyloxy-5-bromopyrimidine 95%

Warning

H27648.MDEA 80.2 EUR
H27648.MD H27648.03
2-Benzyloxy-5-bromopyrimidine 95%
2-Benzyloxy-5-bromopyrimidine
Formula: C₁₁H₉BrN₂O
Melting Pt: 100…103 °C
Storage Temperature: Ambient
MDL Number: MFCD07375140
CAS Number: 742058-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR