Methyldiphenylphosphine oxide

Supplier: Molekula

MOLEM59129780EA 0 EUR
MOLEM59129780
Methyldiphenylphosphine oxide
Methyldiphenylphosphine oxide
Formula: C₁₃H₁₃OP
MW: 216,22 g/mol
MDL Number: MFCD00002081
CAS Number: 2129-89-7
EINECS: 218-346-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR