2-Acetamido-5-nitrothiazole ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: N-(5-Nitro-2-thiazolyl)acetamide

L01356.06EA 191 EUR
L01356.06 L01356.03
2-Acetamido-5-nitrothiazole ≥98%
2-Acetamidot-5-nitrohiazole
Formula: C₅H₅N₃O₃S
MW: 187,18 g/mol
Melting Pt: ∼ 263 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00022438
CAS Number: 140-40-9
EINECS: 205-414-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR