D-(-)-Penicillamine

Supplier: Molekula

Synonyms: H-D-Pen-OH(S)-2-Amino-3-mercapto-3-methylbutanoic acid

MOLE47221894-5GEA 145 EUR
MOLE47221894-5G
D-(-)-Penicillamine
D(-)-Penicillamine
Formula: C₅H₁₁NO₂S
MW: 149,2 g/mol
MDL Number: MFCD00064302
CAS Number: 52-67-5
EINECS: 200-148-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR