3-Trimethylsilyl-1-propylboronic acid pinacol ester ≥94%

Supplier: Thermo Scientific

H28413.06EA 680 EUR
H28413.06 H28413.MD H28413.03
3-Trimethylsilyl-1-propylboronic acid pinacol ester ≥94%
3-Trimethylsilyl-1-propylboronic acid pinacol ester
Formula: C₁₂H₂₇BO₂Si
Storage Temperature: Ambient
MDL Number: MFCD09953502
CAS Number: 165904-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR