4'-(Trifluoromethoxy)acetanilide 98%

Danger

H31762.06EA 19.2 EUR
H31762.06
4'-(Trifluoromethoxy)acetanilide 98%
4'-(Trifluoromethoxy)acetanilide
Formula: C₉H₈F₃NO₂
Melting Pt: 114…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00075293
CAS Number: 1737-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR