1-Pentylmagnesium chloride 1 M in MeTHF (2-methyltetrahydrofuran)

Supplier: Thermo Scientific
Danger

Synonyms: Pentylmagnesium chloride

H51175.APEA 272 EUR
H51175.AP H51175.AE
1-Pentylmagnesium chloride 1 M in MeTHF (2-methyltetrahydrofuran)
1-Pentylmagnesium chloride
Formula: C₅H₁₁ClMg
MW: 130,9
Storage Temperature: Ambient
MDL Number: MFCD00672089
UN: 2924
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR