3-Amino-3-(4-bromophenyl)propionic acid, 97+%

H52372.06EA 676.9 EUR
ALFAH52372.06 ALFAH52372.MD ALFAH52372.03
3-Amino-3-(4-bromophenyl)propionic acid, 97+%
Purity: 97+%
CAS No.: 39773-47-2
Molecular Formula: C9H10BrNO2
Formula Weight: 244.09
MDL No.: MFCD00187112
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR