2'-Benzoyl-2,4'-dichloroacetanilide

Supplier: LGC Standards PROMOCHEM

Synonyms: N1-(2-Benzoyl-4-chlorophenyl)-2-chloroacetamide

PROOMM0023.05EA 1290 EUR
PROOMM0023.05
2'-Benzoyl-2,4'-dichloroacetanilide
2'-Benzoyl-2,4'-dichloroacetanilide
Formula: C₁₅H₁₁Cl₂NO₂
MDL Number: MFCD00018905
CAS Number: 4016-85-7
EINECS: 223-678-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR