3-(3-Fluoro-4-methoxybenzoyl)propionic acid ≥99%

Supplier: Thermo Scientific
Warning

L07042.03EA 36.3 EUR
L07042.03 L07042.06
3-(3-Fluoro-4-methoxybenzoyl)propionic acid ≥99%
3-(3-Fluoro-4-methoxybenzoyl)propionic acid
Formula: C₁₁H₁₁FO₄
MW: 226,20 g/mol
Melting Pt: 169…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00020538
CAS Number: 347-63-7
EINECS: 206-474-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR