Potassium-4-formylphenyltrifluoroborate ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Potassium trifluoro(4-formylphenyl)borate
L17969.06EA
197
EUR
L17969.06
L17969.03
Potassium-4-formylphenyltrifluoroborate ≥97%
Potassium 4-formylphenyltrifluoroborate
Formula:
C₇H₅BF₃KO MW: 212,02 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093980 CAS Number: 374564-36-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...