Potassium-2-(methylthio)phenyltrifluoroborate 96%
|
Warning
|
Synonyms:
Potassium(2-Methylsulphonylphenyl)trifluoroboroate
, Potassium trifluoro(2-(methylthio)phenyl)borate
H52587.06EA
264
EUR
H52587.06
Potassium-2-(methylthio)phenyltrifluoroborate 96%
Potassium 2-(methylthio)phenyltrifluoroborate
Formula:
C₇H₇BF₃KS MW: 230,10 g/mol Melting Pt: 274…276 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115773 CAS Number: 850623-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...