5-Chloro-2-hydroxybenzhydrazide

MAYBRH01278.1EA 0 EUR
CHMPRH01278.250 MAYBRH01278.1
5-Chloro-2-hydroxybenzhydrazide
5-Chloro-2-hydroxybenzhydrazide
Formula: C₇H₇ClN₂O₂
MDL Number: MFCD01001347
CAS Number: 5022-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR