2,6-Dichloro-4-(trifluoromethoxy)benzoic acid ≥97%

Supplier: Alfa Aesar
Warning

H26867.06EA 246 EUR
H26867.06 H26867.03
2,6-Dichloro-4-(trifluoromethoxy)benzoic acid ≥97%
2,6-Dichloro-4-(trifluoromethoxy)benzoic acid
Formula: C₈H₃Cl₂F₃O₃
Melting Pt: 151…153 °C
Storage Temperature: Ambient
MDL Number: MFCD06660297
CAS Number: 886502-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR