3-Bromo-5-(trifluoromethoxy)benzoic acid ≥97%

Supplier: Thermo Scientific
Warning

H26500.03EA 59.1 EUR
H26500.03 H26500.06
3-Bromo-5-(trifluoromethoxy)benzoic acid ≥97%
3-Bromo-5-(trifluoromethoxy)benzoic acid
Formula: C₈H₄BrF₃O₃
Melting Pt: 87…89 °C
Storage Temperature: Ambient
MDL Number: MFCD04116041
CAS Number: 453565-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR