3-Chloro-5-(trifluoromethoxy)benzoic acid ≥97%

Supplier: Alfa Aesar
Warning

H26279.03EA 73.9 EUR
H26279.03 H26279.06
3-Chloro-5-(trifluoromethoxy)benzoic acid ≥97%
3-Chloro-5-(trifluoromethoxy)benzoic acid
Formula: C₈H₄ClF₃O₃
Melting Pt: 64…68 °C
Storage Temperature: Ambient
MDL Number: MFCD06660306
CAS Number: 433926-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR