4-Chloro-2-(trifluoromethoxy)benzoic acid ≥97%
|
Warning
|
H33403.03EA
110
EUR
H33403.03
H33403.06
4-Chloro-2-(trifluoromethoxy)benzoic acid ≥97%
4-Chloro-2-(trifluoromethoxy)benzoic acid
Formula:
C₈H₄ClF₃O₃ MW: 240,56 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18393801 CAS Number: 1261605-70-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...