3-Amino-4-hydroxybenzhydrazide 98%
Supplier: Thermo Scientific
|
Warning
|
A11538.03EA
36.8
EUR
A11538.03
A11538.06
3-Amino-4-hydroxybenzhydrazide 98%
3-Amino-4-hydroxybenzhydrazide
Formula:
C₇H₉N₃O₂ MW: 167,17 g/mol Melting Pt: 250…252 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017068 CAS Number: 7450-57-9 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...