3,4-Diaminobenzhydrazide 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3,4-Diaminobenzohydrazide
L05263.03EA
64.8
EUR
L05263.03
3,4-Diaminobenzhydrazide 98%
3,4-Diaminobenzhydrazide
Formula:
C₇H₁₀N₄O MW: 166,18 g/mol Melting Pt: 148…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017069 CAS Number: 103956-09-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...