Methyl-4-aminosalicylate

Supplier: LGC Standards PROMOCHEM

Synonyms: Methyl 4-amino-2-hydroxybenzoate

PROOMM0073.09EA 1240 EUR
PROOMM0073.09
Methyl-4-aminosalicylate
Methyl 4-aminosalicylate
Formula: C₈H₉NO₃
MW: 167,18
MDL Number: MFCD00088091
CAS Number: 4136-97-4
EINECS: 223-958-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR