2-Hydroxy-5-(trifluoromethoxy)benzonitrile ≥95%

Supplier: Thermo Scientific

Danger

H26101.03EA 176 EUR
H26101.03 H26101.MD
2-Hydroxy-5-(trifluoromethoxy)benzonitrile ≥95%
2-Hydroxy-5-(trifluoromethoxy)benzonitrile
Formula: C₈H₄F₃NO₂
Melting Pt: 84…90 °C
Storage Temperature: Ambient
MDL Number: MFCD04973781
CAS Number: 875664-40-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR