N-Cyclopentyl-4-fluorobenzamide, 97%

H57944.03EA 160.3 EUR
ALFAH57944.03 ALFAH57944.MD
N-Cyclopentyl-4-fluorobenzamide, 97%
Purity: 97%
CAS No.: 300829-28-1
Molecular Formula: C12H14FNO
Formula Weight: 207.25
MDL No.: MFCD00707303
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR