5-(Trifluoromethoxy)salicylic acid ≥98%

Warning

Synonyms: 2-Hydroxy-5-(trifluoromethoxy)benzoic acid

H26099.MDEA 138 EUR
H26099.MD H26099.03
5-(Trifluoromethoxy)salicylic acid ≥98%
5-(Trifluoromethoxy)salicylic acid
Formula: C₈H₅F₃O₄
MW: 222,12
Melting Pt: 131…133 °C
Storage Temperature: Ambient
MDL Number: MFCD01091003
CAS Number: 129644-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR