3-Nitrophenylsulphur pentafluoride 95%

Supplier: Thermo Scientific
Danger

L17545.03EA 45.3 EUR
L17545.03
3-Nitrophenylsulphur pentafluoride 95%
3-Nitrophenylsulphur pentafluoride
Formula: C₆H₄F₅NO₂S
MW: 249,16 g/mol
Boiling Pt: 106 °C (2 mmHg)
Density: 1,662
Storage Temperature: Ambient
MDL Number: MFCD00221621
CAS Number: 2613-26-5
EINECS: 447-600-2
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR