4-Methyl-2-nitrobenzeneboronic acid, 95%

H53181.MDEA 91.2 EUR
ALFAH53181.MD
4-Methyl-2-nitrobenzeneboronic acid, 95%
Purity: 95%
CAS No.: 143697-03-4
Molecular Formula: C7H8BNO4
Formula Weight: 180.96
Melting Point: 240-244°
MDL No.: MFCD06659821
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR