3-(Hydroxymethyl)phenylboronic acid

Supplier: Molekula

Synonyms: 3-(Hydroxymethyl)benzeneboronic acid

MOLE59697211-1GEA 0 EUR
MOLE59697211-1G
3-(Hydroxymethyl)phenylboronic acid
3-(Hydroxymethyl)phenylboronic acid
Formula: C₇H₉BO₃
MW: 151,96
MDL Number: MFCD01317846
CAS Number: 87199-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR