4-(Hydroxymethyl)phenylboronic acid
Supplier: Molekula
Synonyms:
4-(Hydroxymethyl)benzeneboronic acid
MOLEM54140413EA
0
EUR
MOLEM54140413
4-(Hydroxymethyl)phenylboronic acid
4-(Hydroxymethyl)phenylboronic acid
Formula:
C₇H₉BO₃ MW: 151,96 g/mol |
MDL Number:
MFCD00792672 CAS Number: 59016-93-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...