2-Fluoro-N-(3-fluorobenzyl)-5-methylaniline, 97%

H57520.03EA 92.8 EUR
ALFAH57520.03 ALFAH57520.MD
2-Fluoro-N-(3-fluorobenzyl)-5-methylaniline, 97%
Purity: 97%
CAS No.: 1039824-92-4
Molecular Formula: C14H13F2N
Formula Weight: 233.26
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR