3-Fluoro-N-(2-fluorobenzyl)-4-methylaniline, 97%

H57371.MDEA 72.3 EUR
ALFAH57371.MD ALFAH57371.03
3-Fluoro-N-(2-fluorobenzyl)-4-methylaniline, 97%
Purity: 97%
CAS No.: 1021077-25-7
Molecular Formula: C14H13F2N
Formula Weight: 233.26
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR