Acetophenone azine ≥97%
Supplier: Thermo Scientific
Synonyms:
1,2-Bis(1-phenylethylidene)hydrazine
L03924.06EA
185
EUR
L03924.06
Acetophenone azine ≥97%
Acetophenone azine
Formula:
C₁₆H₁₆N₂ MW: 236,32 g/mol Melting Pt: 120…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015011 CAS Number: 729-43-1 EINECS: 211-979-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...