Vanillin azine, Sigma-Aldrich®
235628-5GEA
171
EUR
235628-5G
Vanillin azine, Sigma-Aldrich®
Vanillin azine
Formula:
C₁₆H₁₆N₂O₄ |
MDL Number:
MFCD00008364 CAS Number: 1696-60-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...