3,5-Dimethyl-4-ethoxyphenylboronic acid ≥98%
|
Warning
|
Synonyms:
3,5-Dimethyl-4-ethoxybenzeneboronic acid
, 4-Ethoxy-3,5-dimethylbenzeneboronic acid
, 4-Ethoxy-3,5-dimethylphenylboronic acid
, (4-Ethoxy-3,5-dimethylphenyl)boronic acid
H52767.06EA
126
EUR
H52767.06
3,5-Dimethyl-4-ethoxyphenylboronic acid ≥98%
3,5-Dimethyl-4-ethoxyphenylboronic acid
Formula:
C₁₀H₁₅BO₃ MW: 194,04 g/mol Melting Pt: 210…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179463 CAS Number: 850568-59-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...