3,5-Dimethyl-4-methoxyphenylboronic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-Methoxy-3,5-dimethylbenzeneboronic acid
, 4-Methoxy-3,5-dimethylphenylboronic acid
L19820.03EA
42.7
EUR
L19820.03
3,5-Dimethyl-4-methoxyphenylboronic acid ≥98%
3,5-Dimethyl-4-methoxyphenylboronic acid
Formula:
C₉H₁₃BO₃ MW: 180,01 g/mol Melting Pt: ∼ 237 °C Storage Temperature: Ambient |
MDL Number:
MFCD01114648 CAS Number: 301699-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...